Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01328012_m1
          See other CELF1 GE Assays ›
          Gene Symbol:
          Celf1
          Gene Name:
          CUGBP, Elav-like family member 1
          Gene Aliases:
          Cugbp1
          Chromosome Location:
          Chr.3: 79768901 - 79844548 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01328012_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Celf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025421.1 NP_001020592.1 10 - 11 1218 71
          XM_006234513.2 XP_006234575.1 12 - 13 2149 71
          XM_006234514.1 XP_006234576.1 11 - 12 1359 71
          XM_006234516.1 XP_006234578.1 11 - 12 1359 71
          XM_006234518.1 XP_006234580.1 11 - 12 1356 71
          XM_006234519.1 XP_006234581.1 11 - 12 1347 71
          XM_006234520.1 XP_006234582.1 11 - 12 1344 71
          XM_006234521.1 XP_006234583.1 11 - 12 1344 71
          XM_006234522.1 XP_006234584.1 11 - 12 1281 71
          XM_006234523.1 XP_006234585.1 12 - 13 1417 71
          XM_006234524.2 XP_006234586.1 12 - 13 1308 71
          XM_006234526.2 XP_006234588.1 12 - 13 1532 71
          XM_006234527.2 XP_006234589.1 12 - 13 1529 71
          XM_008762008.1 XP_008760230.1 13 - 14 2351 71
          XM_008762009.1 XP_008760231.1 13 - 14 1434 71
          XM_008762010.1 XP_008760232.1 12 - 13 2198 71
          XM_008762011.1 XP_008760233.1 11 - 12 1228 71
          GenBank mRNA BC098012.1 - 10 - 11 1218 7454358 71
          FQ221418.1 - 11 - 12 1350 71
          FQ222063.1 - 11 - 12 1357 71
          FQ234844.1 - 1 - 2 44 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001123.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Celf1

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splice site selection
          spermatid development
          positive regulation of multicellular organism growth
          regulation of RNA splicing
          nucleotide binding
          RNA binding
          mRNA binding
          translation initiation factor binding
          BRE binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P075515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169408 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075515 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075515 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P075515 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18263 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P396198 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10837765 - -
          Affymetrix Rat Gene 1.0 ST Array 10837756 - -
          Affymetrix Rat Gene 1.0 ST Array 10837762 - -
          Affymetrix Rat Gene 1.0 ST Array 10837764 - -
          Affymetrix Rat Gene 1.0 ST Array 10837758 - -
          Affymetrix Rat Gene 1.0 ST Array 10837760 - -
          Affymetrix Rat Gene 1.0 ST Array 10837761 - -
          Affymetrix Rat Gene 1.0 ST Array 10837759 - -
          Affymetrix Rat Gene 1.0 ST Array 10837754 - -
          Affymetrix Rat Gene 1.0 ST Array 10837755 - -
          Affymetrix Rat Gene 1.0 ST Array 10837763 - -
          Affymetrix Rat Gene 1.0 ST Array 10837757 - -
          Affymetrix Rat Gene 1.0 ST Array 10837766 - -
          Affymetrix Rat Gene 1.1 ST Array 10837752 - -
          Rat Genome 230 2.0 Array 1388704_at - -
          Rat Genome 230 2.0 Array 1393568_at - -
          Rat Genome 230 2.0 Array 1388704_PM_at - -
          Rat Genome 230 2.0 Array 1393568_PM_at - -
          Rat Genome 230 2.0 Array 1371900_at - -
          Rat Genome 230 2.0 Array 1371900_PM_at - -
          Rat Genome 230 2.0 Array 1393257_PM_at - -
          Rat Genome 230 2.0 Array 1393257_at - -
          Affymetrix HT Rat Focus Array 1393568_at - -
          Affymetrix HT Rat Focus Array 1393257_at - -
          Affymetrix HT Rat Focus Array 1393568_PM_at - -
          Affymetrix HT Rat Focus Array 1393257_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300007803.rn.1 TC0300001123.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300007804.rn.1 TC0300001123.rn.1 RTA TC
          Rat Expression Set 230 1371900_PM_at - -
          Rat Expression Set 230 1371900_at - -
          Rat Expression Set 230 1393257_PM_at - -
          Rat Expression Set 230 1393568_at - -
          Rat Expression Set 230 1393568_PM_at - -
          Rat Expression Set 230 1393257_at - -
          Affymetrix Rat Gene 1.0 ST Array 10837752 - -
          Rat Genome U34 Set rc_AA892228_at - -
          Rat Genome U34 Set rc_AI029156_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21992835 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369085 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline