Search
Search
| Gene Symbol: Ncor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271103.1 | NP_001258032.1 | 44 - 45 | 7282 | 62 | |
| GenBank mRNA | AF124821.1 | - | 9 - 10 | 1410 | 62 | |
| FQ225703.1 | - | 1 - 2 | 156 | 62 | ||
| FQ233813.1 | - | 1 - 2 | 156 | 62 | ||
Set Membership: |
|