Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01399560_m1
          See other RXRB GE Assays ›
          Gene Symbol:
          Rxrb
          Gene Name:
          retinoid X receptor beta
          Gene Aliases:
          RXR-beta
          Chromosome Location:
          Chr.20: 3822678 - 3829138 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01399560_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rxrb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206849.3 NP_996731.2 4 - 5 779 70
          XM_006255960.2 XP_006256022.1 4 - 5 1289 70
          XM_006255961.2 XP_006256023.1 4 - 5 782 70
          GenBank mRNA BC099776.1 - 4 - 5 681 70
          M81766.1 - 4 - 5 608 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000125.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rxrb

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          maternal placenta development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          regulation of myelination
          steroid hormone mediated signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          retinoic acid receptor signaling pathway
          ventricular cardiac muscle cell differentiation
          cardiac muscle cell proliferation
          cellular response to retinoic acid
          positive regulation of receptor activity
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          9-cis retinoic acid receptor activity
          zinc ion binding
          thyroid hormone receptor coactivator activity
          protein homodimerization activity
          vitamin D receptor binding
          retinoic acid receptor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          thyroid hormone receptor binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P327151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9732 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113744 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P327151 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P327151 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P327151 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P402675 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700479934 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700767450 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54276 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18146 - -
          Agilent Rat cDNA Microarray (G4105A) 700479934 - -
          Agilent Rat cDNA Microarray (G4105A) 700767450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10831716 - -
          Affymetrix Rat Gene 1.0 ST Array 10831723 - -
          Affymetrix Rat Gene 1.0 ST Array 10831717 - -
          Affymetrix Rat Gene 1.0 ST Array 10831722 - -
          Affymetrix Rat Gene 1.0 ST Array 10831721 - -
          Affymetrix Rat Gene 1.0 ST Array 10831718 - -
          Affymetrix Rat Gene 1.0 ST Array 10831720 - -
          Affymetrix Rat Gene 1.0 ST Array 10831725 - -
          Affymetrix Rat Gene 1.0 ST Array 10831719 - -
          Affymetrix Rat Gene 1.1 ST Array 10831715 - -
          Rat Genome 230 2.0 Array 1389051_PM_at - -
          Rat Genome 230 2.0 Array 1389051_at - -
          Affymetrix HT Rat Focus Array 1389051_at - -
          Affymetrix HT Rat Focus Array 1389051_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000000906.rn.1 TC2000000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000907.rn.1 TC2000000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000911.rn.1 TC2000000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000910.rn.1 TC2000000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000915.rn.1 TC2000000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000903.rn.1 TC2000000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000914.rn.1 TC2000000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000905.rn.1 TC2000000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000916.rn.1 TC2000000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000908.rn.1 TC2000000125.rn.1 RTA TC
          Rat Expression Set 230 1389051_PM_at - -
          Rat Expression Set 230 1389051_at - -
          Affymetrix Rat Gene 1.0 ST Array 10831715 - -
          Rat Genome U34 Set M81766_at - -
          Rat Genome U34 Set M81766_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21929379 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376944 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375706 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline