Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01399619_m1
          See other MBP GE Assays ›
          Gene Symbol:
          Mbp
          Gene Name:
          myelin basic protein
          Gene Aliases:
          Mbps
          Chromosome Location:
          Chr.18: 79326636 - 79437310 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01399619_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mbp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025291.1 NP_001020462.1 3 - 4 434 80
          NM_001025292.1 NP_001020463.1 2 - 3 356 80
          NM_001025293.1 NP_001020464.1 3 - 4 434 80
          NM_017026.2 NP_058722.1 2 - 3 356 80
          XM_006254997.2 XP_006255059.1 6 - 7 1096 80
          XM_008772082.1 XP_008770304.1 6 - 7 1111 80
          XM_008772083.1 XP_008770305.1 5 - 6 651 80
          GenBank mRNA AJ132895.1 - 2 - 3 279 80
          AJ132896.1 - 3 - 4 357 80
          AJ132897.1 - 2 - 3 279 80
          AJ132898.1 - 3 - 4 357 80
          BC094522.1 - 2 - 3 312 7316167 80
          CB586909.1 - 2 - 3 384 80
          FQ214209.1 - 2 - 3 323 80
          JX131663.1 - 3 - 4 434 80
          K00512.1 - 2 - 3 303 80
          M25889.1 - 2 - 3 319 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000934.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mbp

          Panther Classification:

          Molecular Function -

          myelin protein

          Gene Ontology Categories:

          Function(s) Process(es)

          aging
          response to toxic substance
          substantia nigra development
          response to progesterone
          response to tumor necrosis factor
          myelination
          response to mercury ion
          negative regulation of axonogenesis
          membrane organization
          response to fatty acid
          protease binding
          protein binding
          structural constituent of myelin sheath

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017160 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55584 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62861 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157404 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172020 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96876 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131135 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017163 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017160 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017163 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017160 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017160 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017163 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11782 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P334799 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700256012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71289 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89681 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67890 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17444 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6951 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78727 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66878 - -
          Agilent Rat cDNA Microarray (G4105A) 700256012 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802975 - -
          Affymetrix Rat Gene 1.0 ST Array 10802989 - -
          Affymetrix Rat Gene 1.0 ST Array 10802986 - -
          Affymetrix Rat Gene 1.0 ST Array 10802973 - -
          Affymetrix Rat Gene 1.0 ST Array 10802984 - -
          Affymetrix Rat Gene 1.0 ST Array 10802981 - -
          Affymetrix Rat Gene 1.0 ST Array 10802983 - -
          Affymetrix Rat Gene 1.0 ST Array 10802987 - -
          Affymetrix Rat Gene 1.0 ST Array 10802979 - -
          Affymetrix Rat Gene 1.0 ST Array 10802982 - -
          Affymetrix Rat Gene 1.0 ST Array 10802978 - -
          Affymetrix Rat Gene 1.0 ST Array 10802977 - -
          Affymetrix Rat Gene 1.0 ST Array 10802976 - -
          Affymetrix Rat Gene 1.0 ST Array 10802988 - -
          Affymetrix Rat Gene 1.0 ST Array 10802974 - -
          Affymetrix Rat Gene 1.0 ST Array 10802985 - -
          Affymetrix Rat Gene 1.0 ST Array 10802990 - -
          Affymetrix Rat Gene 1.1 ST Array 10802972 - -
          Affymetrix Rat Gene 1.1 ST Array 10802980 - -
          Rat Genome 230 2.0 Array 1368810_PM_a_at - -
          Rat Genome 230 2.0 Array 1368810_a_at - -
          Rat Genome 230 2.0 Array 1387341_PM_a_at - -
          Rat Genome 230 2.0 Array 1387341_a_at - -
          Affymetrix HT Rat Focus Array 1368810_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387341_a_at - -
          Affymetrix HT Rat Focus Array 1368810_a_at - -
          Affymetrix HT Rat Focus Array 1387341_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1800006627.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006646.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006640.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006616.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006628.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006643.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006621.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006619.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006637.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006639.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006645.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006622.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006642.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006638.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006614.rn.1 TC1800000934.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006624.rn.1 TC1800000934.rn.1 RTA TC
          Rat Expression Set 230 1368810_a_at - -
          Rat Expression Set 230 1368810_PM_a_at - -
          Rat Expression Set 230 1387341_a_at - -
          Rat Expression Set 230 1387341_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802972 - -
          Affymetrix Rat Gene 1.0 ST Array 10802980 - -
          Rat Genome U34 Set K00512_at - -
          Rat Genome U34 Set rc_AI145512_at - -
          Rat Genome U34 Set K00512_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359702 - -
          RatRef-12 Expression BeadChip ILMN_48846 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline