Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01399683_m1
          See other CNDP2 GE Assays ›
          Gene Symbol:
          Cndp2
          Gene Name:
          CNDP dipeptidase 2 (metallopeptidase M20 family)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 81521966 - 81539065 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01399683_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cndp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010920.1 NP_001010920.1 11 - 12 1455 62
          XM_006254998.2 XP_006255060.1 10 - 11 1501 62
          XM_006254999.2 XP_006255061.1 11 - 12 1495 62
          GenBank mRNA AY569013.1 - 11 - 12 1455 62
          BC095904.1 - 10 - 11 1428 7110846 62
          FQ223403.1 - 2 - 3 262 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001828.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cndp2

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          carboxypeptidase activity
          peptidase activity
          metallopeptidase activity
          dipeptidase activity
          tripeptidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P297619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570347 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121501 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116056 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P297619 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P297619 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P297619 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P297619 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34679 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41374 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54571 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805550 - -
          Affymetrix Rat Gene 1.0 ST Array 10805541 - -
          Affymetrix Rat Gene 1.0 ST Array 10805552 - -
          Affymetrix Rat Gene 1.0 ST Array 10805547 - -
          Affymetrix Rat Gene 1.0 ST Array 10805543 - -
          Affymetrix Rat Gene 1.0 ST Array 10805551 - -
          Affymetrix Rat Gene 1.0 ST Array 10805553 - -
          Affymetrix Rat Gene 1.0 ST Array 10805554 - -
          Affymetrix Rat Gene 1.0 ST Array 10805545 - -
          Affymetrix Rat Gene 1.0 ST Array 10805548 - -
          Affymetrix Rat Gene 1.0 ST Array 10805549 - -
          Affymetrix Rat Gene 1.0 ST Array 10805544 - -
          Affymetrix Rat Gene 1.0 ST Array 10805555 - -
          Affymetrix Rat Gene 1.0 ST Array 10805542 - -
          Affymetrix Rat Gene 1.0 ST Array 10805546 - -
          Affymetrix Rat Gene 1.1 ST Array 10805540 - -
          Rat Genome 230 2.0 Array 1372132_at - -
          Rat Genome 230 2.0 Array 1372132_PM_at - -
          Affymetrix HT Rat Focus Array 1372132_PM_at - -
          Affymetrix HT Rat Focus Array 1372132_at - -
          Affymetrix Rat Clariom S Assay PSR1800012763.rn.1 TC1800001828.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800012762.rn.1 TC1800001828.rn.1 RTA TC
          Rat Expression Set 230 1372132_at - -
          Rat Expression Set 230 1372132_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805540 - -
          Rat Genome U34 Set rc_AI043964_s_at - -
          Rat Genome U34 Set H34730_at - -
          Rat Genome U34 Set AA801349_at - -
          Rat Genome U34 Set rc_AI168975_g_at - -
          Rat Genome U34 Set rc_AI168975_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22422125 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376891 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530308 - -
          RatRef-12 Expression BeadChip ILMN_161188 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline