Search
Search
| Gene Symbol: Cbln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012740.1 | NP_001012758.1 | 1 - 2 | 535 | 63 | |
| XM_006255022.2 | XP_006255084.1 | 3 - 4 | 1036 | 63 | ||
| XM_006255023.2 | XP_006255085.1 | 2 - 3 | 991 | 63 | ||
| XM_006255024.2 | XP_006255086.1 | 2 - 3 | 671 | 63 | ||
| GenBank mRNA | BC091341.1 | - | 1 - 2 | 535 | 7315458 | 63 |
Set Membership: |
|