Search
Search
| Gene Symbol: Focad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238379.2 | XP_006238441.1 | 35 - 36 | 4577 | 63 | |
| XM_233144.7 | XP_233144.5 | 41 - 42 | 5102 | 63 | ||
| GenBank mRNA | FQ233601.1 | - | 9 - 10 | 1240 | 63 | |
Set Membership: |
|