Search
Search
| Gene Symbol: Gmppa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025056.1 | NP_001020227.1 | 6 - 7 | 702 | 63 | |
| XM_006245267.2 | XP_006245329.1 | 6 - 7 | 605 | 63 | ||
| XM_006245268.2 | XP_006245330.1 | 6 - 7 | 676 | 63 | ||
| XM_006245269.2 | XP_006245331.1 | 6 - 7 | 670 | 63 | ||
| XM_006245270.2 | XP_006245332.1 | 6 - 7 | 573 | 63 | ||
| XM_006245271.2 | XP_006245333.1 | 6 - 7 | 605 | 63 | ||
| XM_008767291.1 | XP_008765513.1 | 6 - 7 | 712 | 63 | ||
| GenBank mRNA | BC083763.1 | - | 6 - 7 | 702 | 7193128 | 63 |
Set Membership: |
|