Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01400330_m1
          See other SP110 GE Assays ›
          Gene Symbol:
          Sp110
          Gene Name:
          SP110 nuclear body protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 92593909 - 92618808 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01400330_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sp110
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034137.1 NP_001029309.1 12 - 13 1486 126
          XM_006245370.2 XP_006245432.1 11 - 12 1367 126
          XM_008767234.1 XP_008765456.1 13 - 14 1550 126
          GenBank mRNA BC105751.1 - 12 - 13 1486 5599224 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002375.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sp110

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          response to bacterium
          positive regulation of apoptotic process
          innate immune response
          DNA binding
          chromatin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P611638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40552 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91323 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P611638 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P611638 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P611638 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P348614 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19238 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61512 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35069 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53135 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17688 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929542 - -
          Affymetrix Rat Gene 1.0 ST Array 10929555 - -
          Affymetrix Rat Gene 1.0 ST Array 10929546 - -
          Affymetrix Rat Gene 1.0 ST Array 10929547 - -
          Affymetrix Rat Gene 1.0 ST Array 10929550 - -
          Affymetrix Rat Gene 1.0 ST Array 10929551 - -
          Affymetrix Rat Gene 1.0 ST Array 10929544 - -
          Affymetrix Rat Gene 1.0 ST Array 10929543 - -
          Affymetrix Rat Gene 1.0 ST Array 10929545 - -
          Affymetrix Rat Gene 1.0 ST Array 10929549 - -
          Affymetrix Rat Gene 1.0 ST Array 10929554 - -
          Affymetrix Rat Gene 1.0 ST Array 10929552 - -
          Affymetrix Rat Gene 1.0 ST Array 10929553 - -
          Affymetrix Rat Gene 1.0 ST Array 10929548 - -
          Affymetrix Rat Gene 1.1 ST Array 10929541 - -
          Rat Genome 230 2.0 Array 1396268_at - -
          Rat Genome 230 2.0 Array 1396268_PM_at - -
          Rat Genome 230 2.0 Array 1394819_PM_at - -
          Rat Genome 230 2.0 Array 1372930_PM_at - -
          Rat Genome 230 2.0 Array 1394819_at - -
          Rat Genome 230 2.0 Array 1372930_at - -
          Affymetrix HT Rat Focus Array 1396268_PM_at - -
          Affymetrix HT Rat Focus Array 1372930_at - -
          Affymetrix HT Rat Focus Array 1396268_at - -
          Affymetrix HT Rat Focus Array 1394819_at - -
          Affymetrix HT Rat Focus Array 1372930_PM_at - -
          Affymetrix HT Rat Focus Array 1394819_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900019519.rn.1 TC0900002375.rn.1 RTA TC
          Rat Expression Set 230 1372930_at - -
          Rat Expression Set 230 1372930_PM_at - -
          Rat Expression Set 230 1394819_at - -
          Rat Expression Set 230 1396268_at - -
          Rat Expression Set 230 1396268_PM_at - -
          Rat Expression Set 230 1394819_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10929541 - -
          Rat Genome U34 Set rc_AI070313_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21325321 - -
          ABI Rat Genome Survey Microarray 21486606 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350384 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline