Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01400467_m1
          See other SERPINE2 GE Assays ›
          Gene Symbol:
          Serpine2
          Gene Name:
          serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2
          Gene Aliases:
          Gdnpn1, PI-7, Pn-1, Spin4
          Chromosome Location:
          Chr.9: 85560852 - 85626125 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01400467_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpine2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019197.1 NP_062070.1 4 - 5 813 79
          XM_006245161.2 XP_006245223.1 4 - 5 1476 79
          XM_006245162.2 XP_006245224.1 5 - 6 940 79
          XM_008767212.1 XP_008765434.1 4 - 5 800 79
          GenBank mRNA AY780673.1 - 3 - 4 688 79
          FQ228974.1 - 2 - 3 382 79
          M17784.1 - 3 - 4 689 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002315.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          embryo implantation
          blood coagulation
          negative regulation of cell proliferation
          response to radiation
          response to wounding
          negative regulation of peptidase activity
          negative regulation of platelet activation
          negative regulation of plasminogen activation
          negative regulation of sodium ion transport
          negative regulation of endopeptidase activity
          positive regulation of neuron projection development
          negative regulation of phosphatidylinositol 3-kinase signaling
          cerebellar granular layer morphogenesis
          negative regulation of blood coagulation
          negative regulation of cell growth
          secretion by cell
          secretory granule organization
          negative regulation of protein catabolic process
          mating plug formation
          negative regulation of proteolysis
          negative regulation of smoothened signaling pathway
          regulation of timing of cell differentiation
          positive regulation of astrocyte differentiation
          detection of mechanical stimulus involved in sensory perception
          regulation of synaptic transmission, glutamatergic
          long-term synaptic potentiation
          innervation
          seminal vesicle epithelium development
          dense core granule biogenesis
          negative regulation of platelet aggregation
          serine-type endopeptidase inhibitor activity
          receptor binding
          glycosaminoglycan binding
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P438863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4809 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15648 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129116 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22860 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P438863 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P438863 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P438863 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P438863 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409694 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700312223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4019 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68886 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63371 - -
          Agilent Rat cDNA Microarray (G4105A) 700312223 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929289 - -
          Affymetrix Rat Gene 1.0 ST Array 10929292 - -
          Affymetrix Rat Gene 1.0 ST Array 10929297 - -
          Affymetrix Rat Gene 1.0 ST Array 10929296 - -
          Affymetrix Rat Gene 1.0 ST Array 10929293 - -
          Affymetrix Rat Gene 1.0 ST Array 10929295 - -
          Affymetrix Rat Gene 1.0 ST Array 10929290 - -
          Affymetrix Rat Gene 1.0 ST Array 10929294 - -
          Affymetrix Rat Gene 1.0 ST Array 10929291 - -
          Affymetrix Rat Gene 1.1 ST Array 10929288 - -
          Rat Genome 230 2.0 Array 1372440_PM_at - -
          Rat Genome 230 2.0 Array 1372440_at - -
          Affymetrix HT Rat Focus Array 1372440_at - -
          Affymetrix HT Rat Focus Array 1372440_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900018976.rn.1 TC0900002315.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10929288 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21074342 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370763 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline