Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01400677_m1
          See other SPHKAP GE Assays ›
          Gene Symbol:
          Sphkap
          Gene Name:
          SPHK1 interactor, AKAP domain containing
          Gene Aliases:
          RGD1311951, Skip
          Chromosome Location:
          Chr.9: 89050405 - 89195445 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01400677_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sphkap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127492.1 NP_001120964.1 5 - 6 524 57
          XM_006245230.2 XP_006245292.1 5 - 6 696 57
          XM_006245231.2 XP_006245293.1 5 - 6 1097 57
          XM_006245233.2 XP_006245295.1 4 - 5 616 57
          XM_006245234.2 XP_006245296.1 5 - 6 695 57
          XM_008767251.1 XP_008765473.1 4 - 5 1018 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002351.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein kinase A signaling
          protein kinase A binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P122624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26024 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21362 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78815 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78197 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79284 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92832 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115980 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147217 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122624 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122624 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P122624 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P199508 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929463 - -
          Affymetrix Rat Gene 1.0 ST Array 10929467 - -
          Affymetrix Rat Gene 1.0 ST Array 10929470 - -
          Affymetrix Rat Gene 1.0 ST Array 10929460 - -
          Affymetrix Rat Gene 1.0 ST Array 10929465 - -
          Affymetrix Rat Gene 1.0 ST Array 10929464 - -
          Affymetrix Rat Gene 1.0 ST Array 10929468 - -
          Affymetrix Rat Gene 1.0 ST Array 10929469 - -
          Affymetrix Rat Gene 1.0 ST Array 10929461 - -
          Affymetrix Rat Gene 1.0 ST Array 10929462 - -
          Affymetrix Rat Gene 1.0 ST Array 10929471 - -
          Affymetrix Rat Gene 1.0 ST Array 10929459 - -
          Affymetrix Rat Gene 1.0 ST Array 10929472 - -
          Affymetrix Rat Gene 1.1 ST Array 10929458 - -
          Rat Genome 230 2.0 Array 1397828_at - -
          Rat Genome 230 2.0 Array 1374168_PM_at - -
          Rat Genome 230 2.0 Array 1374168_at - -
          Rat Genome 230 2.0 Array 1397828_PM_at - -
          Affymetrix HT Rat Focus Array 1397828_PM_at - -
          Affymetrix HT Rat Focus Array 1397828_at - -
          Affymetrix Rat Clariom S Assay PSR0900019300.rn.1 TC0900002351.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019301.rn.1 TC0900002351.rn.1 RTA TC
          Rat Expression Set 230 1374168_at - -
          Rat Expression Set 230 1374168_PM_at - -
          Rat Expression Set 230 1397828_PM_at - -
          Rat Expression Set 230 1397828_at - -
          Affymetrix Rat Gene 1.0 ST Array 10929458 - -
          Rat Genome U34 Set rc_AA925101_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20907492 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354866 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline