Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01401096_m1
          See other PLCD4 GE Assays ›
          Gene Symbol:
          Plcd4
          Gene Name:
          phospholipase C, delta 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 81814772 - 81858397 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01401096_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plcd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080688.1 NP_542419.1 12 - 13 1897 123
          XM_006245135.2 XP_006245197.1 14 - 15 3068 123
          XM_006245136.2 XP_006245198.1 13 - 14 1909 123
          XM_006245137.2 XP_006245199.1 13 - 14 2208 123
          XM_006245138.2 XP_006245200.1 13 - 14 2083 123
          XM_008767182.1 XP_008765404.1 13 - 14 2487 123
          XM_008767183.1 XP_008765405.1 13 - 14 2487 123
          XM_008767184.1 XP_008765406.1 13 - 14 2526 123
          XM_008767185.1 XP_008765407.1 13 - 14 1910 123
          XM_008767186.1 XP_008765408.1 13 - 14 2488 123
          XM_008767187.1 XP_008765409.1 13 - 14 2485 123
          XM_008767188.1 XP_008765410.1 13 - 14 2433 123
          XM_008767189.1 XP_008765411.1 13 - 14 2487 123
          XM_008767190.1 XP_008765412.1 12 - 13 2390 123
          XM_008767191.1 XP_008765413.1 12 - 13 2387 123
          GenBank mRNA D50455.1 - 12 - 13 1887 123
          U16655.1 - 12 - 13 1897 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000902.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plcd4

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          acrosome reaction
          lipid catabolic process
          intracellular signal transduction
          phosphatidylinositol metabolic process
          phosphatidylinositol phospholipase C activity
          signal transducer activity
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P139485 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123294 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154783 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75324 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059491 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139485 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139485 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059491 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P139485 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059491 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P656837 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701092307 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700777254 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72346 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93600 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47439 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101155 - -
          Agilent Rat cDNA Microarray (G4105A) 701092307 - -
          Agilent Rat cDNA Microarray (G4105A) 700777254 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10924351 - -
          Affymetrix Rat Gene 1.0 ST Array 10924352 - -
          Affymetrix Rat Gene 1.0 ST Array 10924337 - -
          Affymetrix Rat Gene 1.0 ST Array 10924342 - -
          Affymetrix Rat Gene 1.0 ST Array 10924338 - -
          Affymetrix Rat Gene 1.0 ST Array 10924343 - -
          Affymetrix Rat Gene 1.0 ST Array 10924348 - -
          Affymetrix Rat Gene 1.0 ST Array 10924353 - -
          Affymetrix Rat Gene 1.0 ST Array 10924341 - -
          Affymetrix Rat Gene 1.0 ST Array 10924339 - -
          Affymetrix Rat Gene 1.0 ST Array 10924344 - -
          Affymetrix Rat Gene 1.0 ST Array 10924347 - -
          Affymetrix Rat Gene 1.0 ST Array 10924346 - -
          Affymetrix Rat Gene 1.0 ST Array 10924349 - -
          Affymetrix Rat Gene 1.0 ST Array 10924340 - -
          Affymetrix Rat Gene 1.0 ST Array 10924336 - -
          Affymetrix Rat Gene 1.0 ST Array 10924350 - -
          Affymetrix Rat Gene 1.1 ST Array 10924335 - -
          Rat Genome 230 2.0 Array 1387065_PM_at - -
          Rat Genome 230 2.0 Array 1387065_at - -
          Affymetrix HT Rat Focus Array 1387065_PM_at - -
          Affymetrix HT Rat Focus Array 1387065_at - -
          Affymetrix Rat Clariom S Assay PSR0900007476.rn.1 TC0900000902.rn.1 RTA TC
          Rat Expression Set 230 1387065_PM_at - -
          Rat Expression Set 230 1387065_at - -
          Affymetrix Rat Gene 1.0 ST Array 10924335 - -
          Rat Genome U34 Set U16655_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21472527 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366004 - -
          RatRef-12 Expression BeadChip ILMN_62275 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline