Search
Search
| Gene Symbol: Cabyr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143893.1 | NP_001137365.1 | 3 - 4 | 288 | 130 | |
| XM_006254426.2 | XP_006254488.1 | 3 - 4 | 2341 | 130 | ||
| XM_008771955.1 | XP_008770177.1 | 3 - 4 | 2341 | 130 | ||
| GenBank mRNA | BC160837.1 | - | 3 - 4 | 272 | 130 | |
Set Membership: |
|