Search
Search
| Gene Symbol: Aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142366.2 | NP_001135838.1 | 5 | 143 | ||
| NM_001270558.2 | NP_001257487.1 | 4 | 143 | |||
| NM_001270559.2 | NP_001257488.1 | 4 | 143 | |||
| NM_012825.4 | NP_036957.1 | 5 | 143 | |||
Set Membership: |
|