Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01401546_m1
          See other MYL1 GE Assays ›
          Gene Symbol:
          Myl1
          Gene Name:
          myosin, light chain 1
          Gene Aliases:
          MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
          Chromosome Location:
          Chr.9: 73937820 - 73958480 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01401546_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077656.1 NP_001071124.1 5 - 6 657 103
          NM_020104.2 NP_064489.1 5 - 6 475 103
          XM_008767294.1 XP_008765516.1 4 - 5 605 103
          XM_008767295.1 XP_008765517.1 4 - 5 726 103
          GenBank mRNA BI277595.1 - 2 - 3 103
          CR755322.1 - 5 - 6 480 103
          CR756356.1 - 5 - 6 475 103
          FQ214580.1 - 5 - 6 784 103
          FQ214608.1 - 5 - 6 662 103
          FQ214631.1 - 5 - 6 644 103
          FQ214638.1 - 5 - 6 752 103
          FQ214656.1 - 5 - 6 493 103
          FQ214675.1 - 5 - 6 751 103
          FQ214712.1 - 5 - 6 665 103
          FQ214723.1 - 5 - 6 665 103
          FQ214727.1 - 5 - 6 665 103
          FQ214742.1 - 5 - 6 665 103
          FQ214939.1 - 5 - 6 662 103
          FQ215002.1 - 5 - 6 751 103
          FQ215050.1 - 5 - 6 665 103
          FQ215083.1 - 5 - 6 662 103
          FQ215086.1 - 5 - 6 495 103
          FQ215201.1 - 5 - 6 662 103
          FQ215229.1 - 5 - 6 756 103
          FQ215325.1 - 5 - 6 751 103
          FQ215340.1 - 5 - 6 103
          FQ215418.1 - 5 - 6 751 103
          FQ215435.1 - 5 - 6 662 103
          FQ215535.1 - 4 - 5 726 103
          FQ215557.1 - 5 - 6 751 103
          FQ215563.1 - 5 - 6 659 103
          FQ215578.1 - 5 - 6 751 103
          FQ215610.1 - 5 - 6 665 103
          FQ215748.1 - 4 - 5 724 103
          FQ215766.1 - 5 - 6 751 103
          FQ215774.1 - 5 - 6 662 103
          FQ215848.1 - 5 - 6 662 103
          FQ215875.1 - 5 - 6 662 103
          FQ215911.1 - 5 - 6 756 103
          FQ215933.1 - 5 - 6 665 103
          FQ216034.1 - 5 - 6 665 103
          FQ216060.1 - 5 - 6 751 103
          FQ216109.1 - 5 - 6 662 103
          FQ216124.1 - 5 - 6 756 103
          FQ216181.1 - 5 - 6 103
          FQ216192.1 - 5 - 6 751 103
          FQ216213.1 - 6 - 7 659 103
          FQ216218.1 - 5 - 6 662 103
          FQ216229.1 - 5 - 6 932 103
          FQ216238.1 - 5 - 6 751 103
          FQ216308.1 - 5 - 6 493 103
          FQ216407.1 - 5 - 6 833 103
          FQ216455.1 - 5 - 6 666 103
          FQ216484.1 - 5 - 6 644 103
          FQ216496.1 - 5 - 6 665 103
          FQ216525.1 - 5 - 6 665 103
          FQ216543.1 - 5 - 6 662 103
          FQ216556.1 - 5 - 6 751 103
          FQ216567.1 - 5 - 6 662 103
          FQ216631.1 - 5 - 6 665 103
          FQ216654.1 - 5 - 6 749 103
          FQ216704.1 - 5 - 6 665 103
          FQ216737.1 - 5 - 6 748 103
          FQ217074.1 - 5 - 6 492 103
          FQ217103.1 - 5 - 6 493 103
          FQ217157.1 - 4 - 5 468 103
          FQ217192.1 - 5 - 6 493 103
          FQ217218.1 - 5 - 6 493 103
          FQ217243.1 - 5 - 6 492 103
          FQ217303.1 - 5 - 6 644 103
          FQ217323.1 - 5 - 6 493 103
          FQ217387.1 - 5 - 6 493 103
          FQ217394.1 - 5 - 6 595 103
          FQ217481.1 - 5 - 6 493 103
          FQ217524.1 - 5 - 6 493 103
          FQ217753.1 - 5 - 6 498 103
          FQ217756.1 - 5 - 6 493 103
          FQ217889.1 - 5 - 6 493 103
          FQ217979.1 - 5 - 6 493 103
          FQ218029.1 - 5 - 6 493 103
          FQ218049.1 - 5 - 6 490 103
          FQ223584.1 - 5 - 6 751 103
          FQ223632.1 - 5 - 6 665 103
          FQ223641.1 - 5 - 6 751 103
          FQ223687.1 - 5 - 6 493 103
          FQ223755.1 - 5 - 6 661 103
          FQ223779.1 - 5 - 6 493 103
          FQ223827.1 - 5 - 6 665 103
          FQ223880.1 - 5 - 6 563 103
          FQ223949.1 - 5 - 6 491 103
          FQ223984.1 - 5 - 6 493 103
          FQ224047.1 - 5 - 6 493 103
          FQ224087.1 - 5 - 6 493 103
          FQ224112.1 - 4 - 5 468 103
          FQ224155.1 - 5 - 6 498 103
          FQ224171.1 - 5 - 6 662 103
          FQ224172.1 - 5 - 6 493 103
          FQ224212.1 - 5 - 6 645 103
          FQ224277.1 - 5 - 6 665 103
          FQ224321.1 - 5 - 6 497 103
          FQ224386.1 - 5 - 6 498 103
          FQ224411.1 - 5 - 6 493 103
          FQ224420.1 - 5 - 6 493 103
          FQ224609.1 - 6 - 7 521 103
          FQ224645.1 - 2 - 3 93 103
          FQ224663.1 - 5 - 6 493 103
          FQ224726.1 - 5 - 6 595 103
          FQ224799.1 - 5 - 6 498 103
          FQ224927.1 - 5 - 6 751 103
          FQ224931.1 - 5 - 6 662 103
          FQ224934.1 - 5 - 6 660 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002140.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myl1

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          cardiac muscle contraction
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P531653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119073 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P531653 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P531653 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P531653 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P531653 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P505274 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18265 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91812 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10928625 - -
          Affymetrix Rat Gene 1.0 ST Array 10928618 - -
          Affymetrix Rat Gene 1.0 ST Array 10928623 - -
          Affymetrix Rat Gene 1.0 ST Array 10928616 - -
          Affymetrix Rat Gene 1.0 ST Array 10928624 - -
          Affymetrix Rat Gene 1.0 ST Array 10928620 - -
          Affymetrix Rat Gene 1.0 ST Array 10928622 - -
          Affymetrix Rat Gene 1.0 ST Array 10928621 - -
          Affymetrix Rat Gene 1.0 ST Array 10928619 - -
          Affymetrix Rat Gene 1.0 ST Array 10928615 - -
          Affymetrix Rat Gene 1.1 ST Array 10928614 - -
          Rat Genome 230 2.0 Array 1370033_PM_at - -
          Rat Genome 230 2.0 Array 1370033_at - -
          Affymetrix HT Rat Focus Array 1370033_PM_at - -
          Affymetrix HT Rat Focus Array 1370033_at - -
          Affymetrix Rat Clariom S Assay PSR0900017597.rn.1 TC0900002140.rn.1 RTA TC
          Rat Expression Set 230 1370033_PM_at - -
          Rat Expression Set 230 1370033_at - -
          Affymetrix Rat Gene 1.0 ST Array 10928614 - -
          Rat Genome U34 Set K02423cds_s_at - -
          Rat Genome U34 Set rc_AI170985_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22133845 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361866 - -
          RatRef-12 Expression BeadChip ILMN_66554 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline