Search
Search
| Gene Symbol: Zbtb45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107478.1 | NP_001100948.1 | 2 - 3 | 1499 | 83 | |
| XM_006228114.2 | XP_006228176.1 | 2 - 3 | 2760 | 83 | ||
| XM_006228115.2 | XP_006228177.1 | 2 - 3 | 2668 | 83 | ||
Set Membership: |
|