Search
Search
| Gene Symbol: Zfp606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228138.2 | XP_006228200.1 | 6 - 7 | 475 | 78 | |
| XM_006228139.2 | XP_006228201.1 | 5 - 6 | 422 | 78 | ||
| XM_218283.7 | XP_218283.5 | 5 - 6 | 439 | 78 | ||
Set Membership: |
|