Search
Search

Pds5b| Gene Symbol: Pds5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101805.1 | NP_001095275.1 | 32 - 33 | 4217 | 69 | |
| NM_001102383.2 | NP_001095853.1 | 34 - 35 | 4328 | 69 | ||
| XM_008768959.1 | XP_008767181.1 | 34 - 35 | 4348 | 69 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P991114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158181 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P991114 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P991114 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P991114 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P991114 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P247408 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P991121 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P547976 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P247411 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41371 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75931 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51155 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43978 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10759774 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759781 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759803 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759799 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759777 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759778 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759791 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759773 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759770 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759802 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759771 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759795 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759801 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759776 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759780 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759779 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759800 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759789 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759775 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759782 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759796 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759787 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759797 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759798 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759794 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759804 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759772 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759788 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10759769 | - | - |
| Rat Genome 230 2.0 Array | 1392891_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377100_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392891_at | - | - |
| Rat Genome 230 2.0 Array | 1395053_at | - | - |
| Rat Genome 230 2.0 Array | 1377100_at | - | - |
| Rat Genome 230 2.0 Array | 1395053_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377100_at | - | - |
| Affymetrix HT Rat Focus Array | 1392891_at | - | - |
| Affymetrix HT Rat Focus Array | 1395053_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377100_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392891_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395053_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200000063.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000070.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000084.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000100.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000104.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000058.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000059.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000066.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000067.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000068.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000062.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000064.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000077.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000078.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000106.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000060.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000072.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000073.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000057.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000075.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000076.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000079.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000082.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000054.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000061.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000065.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000071.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000081.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000098.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000069.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000074.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000096.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000055.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000056.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000080.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000097.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000103.rn.1 | TC1200000008.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200000105.rn.1 | TC1200000008.rn.1 | RTA TC |
| Rat Expression Set 230 | 1377100_PM_at | - | - |
| Rat Expression Set 230 | 1377100_at | - | - |
| Rat Expression Set 230 | 1395053_at | - | - |
| Rat Expression Set 230 | 1392891_PM_at | - | - |
| Rat Expression Set 230 | 1395053_PM_at | - | - |
| Rat Expression Set 230 | 1392891_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759769 | - | - |
| Rat Genome U34 Set | rc_AI071670_at | - | - |
| Rat Genome U34 Set | rc_AI229206_at | - | - |
| Rat Genome U34 Set | rc_AI172172_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21309349 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367145 | - | - |