Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01402347_m1
See other MAP3K2 GE Assays ›
Gene Symbol:
Map3k2
Gene Name:
mitogen activated protein kinase kinase kinase 2
Gene Aliases:
Mekk2
Chromosome Location:
Chr.18: 24961250 - 25025488 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01402347_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Map3k2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_138503.1
NP_612512.1
9 - 10
1184
62
GenBank mRNA
AF487544.1
-
2 - 3
129
62
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1800000230.rn.1
Set Membership:
Amplicon length less than or equal to 70
Protein Kinase
Recommended Assay
Rattus norvegicus
1700
Inventoried
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
activation of MAPK activity
protein phosphorylation
cell proliferation
positive regulation of transcription, DNA-templated
cellular response to mechanical stimulus
protein kinase activity
MAP kinase kinase activity
MAP kinase kinase kinase activity
ATP binding
protein kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_64_P071139
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P071139
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P071139
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_64_P071139
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_44_P407402
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_44_P531876
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Gene 1.0 ST Array
10800734
-
-
Affymetrix Rat Gene 1.0 ST Array
10800733
-
-
Affymetrix Rat Gene 1.1 ST Array
10800726
-
-
Affymetrix Rat Clariom S Assay
PSR1800001585.rn.1
TC1800000230.rn.1
RTA TC
Affymetrix Rat Gene 1.0 ST Array
10800726
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
21751674
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline