Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01402349_s1
See other PCDHB2 GE Assays ›
Gene Symbol:
Pcdhb2
Gene Name:
protocadherin beta 2
Gene Aliases:
RGD1563132
Chromosome Location:
Chr.18: 30375798 - 30378194 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01402349_s1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Pcdhb2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001109123.1
NP_001102593.1
1
1042
83
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact
Tech Support
if you need more information.
Set Membership:
Amplicon length between 71 and 85
3' Most
Detect Genomic DNA
Recommended Assay
Rattus norvegicus
1700
Both primers and probe map within a single exon
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_64_P139028
-
-
Agilent SurePrint G3 Rat CGH Microarray 1x1M
190859
-
-
Agilent SurePrint G3 Rat CGH Microarray 1x1M
275679
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P139029
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P139029
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_64_P139029
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Gene 1.0 ST Array
10801169
-
-
Affymetrix Rat Gene 1.1 ST Array
10801167
-
-
Affymetrix Rat Clariom S Assay
PSR1800002593.rn.1
TC1800000327.rn.1
RTA TC
Affymetrix Rat Gene 1.0 ST Array
10801167
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
22313949
-
-
Array Name
Probe IDs
Targets
Target Type
Illumina ratRef-12 v1.0 expression beadchip
ILMN_1651150
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline