Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402419_g1
          See other SLC1A3 GE Assays ›
          Gene Symbol:
          Slc1a3
          Gene Name:
          solute carrier family 1 (glial high affinity glutamate transporter), member 3
          Gene Aliases:
          EAAT1, GLAST, GLAST-1, GluT-1
          Chromosome Location:
          Chr.2: 57860881 - 57935363 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402419_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc1a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289941.1 NP_001276870.1 6 - 7 1234 99
          NM_001289942.1 NP_001276871.1 7 - 8 1361 99
          NM_001289943.1 NP_001276872.1 5 - 6 1025 99
          NM_019225.2 NP_062098.1 7 - 8 1351 99
          GenBank mRNA AF265360.1 - 6 - 7 1073 99
          S59158.1 - 7 - 8 1293 99
          S75687.1 - 3 - 4 577 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003049.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc1a3

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          glutamate metabolic process
          glutamate biosynthetic process
          neurotransmitter transport
          sensory perception of sound
          response to light stimulus
          gamma-aminobutyric acid biosynthetic process
          response to wounding
          response to inorganic substance
          L-glutamate transport
          cranial nerve development
          auditory behavior
          sodium ion transmembrane transport
          response to drug
          malate-aspartate shuttle
          response to antibiotic
          cell morphogenesis involved in neuron differentiation
          positive regulation of synaptic transmission
          neuromuscular process controlling balance
          L-glutamate import
          D-aspartate import
          L-glutamate transmembrane transport
          L-glutamate transmembrane transporter activity
          high-affinity glutamate transmembrane transporter activity
          glutamate binding
          amino acid binding
          sodium:dicarboxylate symporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P113725 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P021516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9585 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108742 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132888 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67330 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021516 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021516 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113725 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P113725 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P021516 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P167719 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P533183 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83318 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77068 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8421 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79271 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34691 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821827 - -
          Affymetrix Rat Gene 1.0 ST Array 10821830 - -
          Affymetrix Rat Gene 1.0 ST Array 10821833 - -
          Affymetrix Rat Gene 1.0 ST Array 10821829 - -
          Affymetrix Rat Gene 1.0 ST Array 10821828 - -
          Affymetrix Rat Gene 1.0 ST Array 10821831 - -
          Affymetrix Rat Gene 1.0 ST Array 10821836 - -
          Affymetrix Rat Gene 1.0 ST Array 10821835 - -
          Affymetrix Rat Gene 1.0 ST Array 10821837 - -
          Affymetrix Rat Gene 1.0 ST Array 10821825 - -
          Affymetrix Rat Gene 1.0 ST Array 10821832 - -
          Affymetrix Rat Gene 1.0 ST Array 10821834 - -
          Affymetrix Rat Gene 1.0 ST Array 10821826 - -
          Affymetrix Rat Gene 1.1 ST Array 10821824 - -
          Rat Genome 230 2.0 Array 1388064_PM_a_at - -
          Rat Genome 230 2.0 Array 1388064_a_at - -
          Rat Genome 230 2.0 Array 1371130_PM_at - -
          Rat Genome 230 2.0 Array 1368565_PM_at - -
          Rat Genome 230 2.0 Array 1368565_at - -
          Rat Genome 230 2.0 Array 1371130_at - -
          Affymetrix HT Rat Focus Array 1368565_PM_at - -
          Affymetrix HT Rat Focus Array 1371130_PM_at - -
          Affymetrix HT Rat Focus Array 1368565_at - -
          Affymetrix HT Rat Focus Array 1371130_at - -
          Affymetrix HT Rat Focus Array 1388064_a_at - -
          Affymetrix HT Rat Focus Array 1388064_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0200021356.rn.1 TC0200003049.rn.1 RTA TC
          Rat Expression Set 230 1371130_at - -
          Rat Expression Set 230 1368565_PM_at - -
          Rat Expression Set 230 1371130_PM_at - -
          Rat Expression Set 230 1368565_at - -
          Rat Expression Set 230 1388064_a_at - -
          Rat Expression Set 230 1388064_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10821824 - -
          Rat Genome U34 Set X63744_at - -
          Rat Genome U34 Set X63744_g_at - -
          Rat Genome U34 Set S75687_s_at - -
          Rat Genome U34 Set S59158_at - -
          Rat Genome U34 Set rc_AA899125_at - -
          Rat Genome U34 Set X63744_g_at - -
          Rat Genome U34 Set X63744_at - -
          Rat Genome U34 Set S59158_at - -
          Rat Genome U34 Set S75687_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363342 - -
          RatRef-12 Expression BeadChip ILMN_66293 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline