Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402476_m1
          See other WDR70 GE Assays ›
          Gene Symbol:
          Wdr70
          Gene Name:
          WD repeat domain 70
          Gene Aliases:
          RGD1309487
          Chromosome Location:
          Chr.2: 56944462 - 57196522 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402476_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wdr70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013909.1 NP_001013931.1 16 - 17 1730 62
          GenBank mRNA BC089903.1 - 16 - 17 1730 7384104 62
          FQ214211.1 - 1 - 2 61 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003035.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wdr70

          Gene Ontology Categories:

          Process(es)

          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P644551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63099 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46534 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7068 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77041 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151971 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140307 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30450 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180768 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98030 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151870 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57251 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114951 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P644551 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P644551 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P644551 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P245864 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P412491 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84297 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6804 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63965 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51290 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102681 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62137 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84641 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93529 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57902 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12909 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35120 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60640 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821761 - -
          Affymetrix Rat Gene 1.0 ST Array 10821764 - -
          Affymetrix Rat Gene 1.0 ST Array 10821756 - -
          Affymetrix Rat Gene 1.0 ST Array 10821752 - -
          Affymetrix Rat Gene 1.0 ST Array 10821768 - -
          Affymetrix Rat Gene 1.0 ST Array 10821753 - -
          Affymetrix Rat Gene 1.0 ST Array 10821759 - -
          Affymetrix Rat Gene 1.0 ST Array 10821758 - -
          Affymetrix Rat Gene 1.0 ST Array 10821766 - -
          Affymetrix Rat Gene 1.0 ST Array 10821754 - -
          Affymetrix Rat Gene 1.0 ST Array 10821757 - -
          Affymetrix Rat Gene 1.0 ST Array 10821760 - -
          Affymetrix Rat Gene 1.0 ST Array 10821767 - -
          Affymetrix Rat Gene 1.0 ST Array 10821750 - -
          Affymetrix Rat Gene 1.0 ST Array 10821751 - -
          Affymetrix Rat Gene 1.0 ST Array 10821755 - -
          Affymetrix Rat Gene 1.0 ST Array 10821762 - -
          Affymetrix Rat Gene 1.0 ST Array 10821763 - -
          Affymetrix Rat Gene 1.1 ST Array 10821749 - -
          Rat Genome 230 2.0 Array 1374715_PM_at - -
          Rat Genome 230 2.0 Array 1374715_at - -
          Affymetrix HT Rat Focus Array 1374715_at - -
          Affymetrix HT Rat Focus Array 1374715_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200021233.rn.1 TC0200003035.rn.1 RTA TC
          Rat Expression Set 230 1374715_at - -
          Rat Expression Set 230 1374715_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10821749 - -
          Rat Genome U34 Set rc_AI012824_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20770771 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353325 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0