Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402599_m1
          See other OSMR GE Assays ›
          Gene Symbol:
          Osmr
          Gene Name:
          oncostatin M receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 56107491 - 56162950 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402599_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Osmr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005384.1 NP_001005384.1 16 - 17 2359 85
          XM_006232012.2 XP_006232074.1 16 - 17 2377 85
          GenBank mRNA AB167522.1 - 16 - 17 2359 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003027.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Osmr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          cytokine-mediated signaling pathway
          response to cytokine
          oncostatin-M-mediated signaling pathway
          cytokine receptor activity
          oncostatin-M receptor activity
          growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37986 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174737 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85752 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018811 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018811 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018811 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P426638 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49511 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38851 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72854 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77131 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821704 - -
          Affymetrix Rat Gene 1.0 ST Array 10821707 - -
          Affymetrix Rat Gene 1.0 ST Array 10821711 - -
          Affymetrix Rat Gene 1.0 ST Array 10821703 - -
          Affymetrix Rat Gene 1.0 ST Array 10821706 - -
          Affymetrix Rat Gene 1.0 ST Array 10821702 - -
          Affymetrix Rat Gene 1.0 ST Array 10821710 - -
          Affymetrix Rat Gene 1.0 ST Array 10821714 - -
          Affymetrix Rat Gene 1.0 ST Array 10821701 - -
          Affymetrix Rat Gene 1.0 ST Array 10821708 - -
          Affymetrix Rat Gene 1.0 ST Array 10821700 - -
          Affymetrix Rat Gene 1.0 ST Array 10821712 - -
          Affymetrix Rat Gene 1.0 ST Array 10821699 - -
          Affymetrix Rat Gene 1.0 ST Array 10821705 - -
          Affymetrix Rat Gene 1.0 ST Array 10821715 - -
          Affymetrix Rat Gene 1.0 ST Array 10821709 - -
          Affymetrix Rat Gene 1.0 ST Array 10821713 - -
          Affymetrix Rat Gene 1.1 ST Array 10821698 - -
          Affymetrix Rat Clariom S Assay PSR0200021163.rn.1 TC0200003027.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10821698 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21577810 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359159 - -
          RatRef-12 Expression BeadChip ILMN_47808 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline