Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402825_m1
          See other TMEM192 GE Assays ›
          Gene Symbol:
          Tmem192
          Gene Name:
          transmembrane protein 192
          Gene Aliases:
          RGD1309341
          Chromosome Location:
          Chr.16: 26620525 - 26642537 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402825_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem192
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014141.1 NP_001014163.1 4 - 5 735 76
          GenBank mRNA BC086402.1 - 4 - 5 735 7304126 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001489.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem192

          Gene Ontology Categories:

          Process(es)

          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P266705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89508 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P266705 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P266705 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P266705 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P266705 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791322 - -
          Affymetrix Rat Gene 1.0 ST Array 10791320 - -
          Affymetrix Rat Gene 1.0 ST Array 10791323 - -
          Affymetrix Rat Gene 1.0 ST Array 10791321 - -
          Affymetrix Rat Gene 1.0 ST Array 10791319 - -
          Affymetrix Rat Gene 1.1 ST Array 10791318 - -
          Rat Genome 230 2.0 Array 1371932_PM_at - -
          Rat Genome 230 2.0 Array 1371932_at - -
          Affymetrix HT Rat Focus Array 1371932_PM_at - -
          Affymetrix HT Rat Focus Array 1371932_at - -
          Affymetrix Rat Clariom S Assay PSR1600011467.rn.1 TC1600001489.rn.1 RTA TC
          Rat Expression Set 230 1371932_PM_at - -
          Rat Expression Set 230 1371932_at - -
          Affymetrix Rat Gene 1.0 ST Array 10791318 - -
          Rat Genome U34 Set rc_AI232287_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21385752 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364059 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530379 - -
          RatRef-12 Expression BeadChip ILMN_161234 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline