Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01403958_m1
          See other NMNAT3 GE Assays ›
          Gene Symbol:
          Nmnat3
          Gene Name:
          nicotinamide nucleotide adenylyltransferase 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 106317043 - 106429535 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01403958_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nmnat3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013224.1 NP_001013242.1 4 - 5 509 72
          XM_006243608.2 XP_006243670.1 4 - 5 853 72
          XM_006243609.2 XP_006243671.1 4 - 5 857 72
          XM_006243610.2 XP_006243672.1 4 - 5 473 72
          GenBank mRNA BC083725.1 - 4 - 5 509 7191168 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001354.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nmnat3

          Panther Classification:

          Molecular Function -

          nucleotidyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          NAD biosynthetic process
          response to wounding
          nicotinamide-nucleotide adenylyltransferase activity
          nicotinate-nucleotide adenylyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105954 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43844 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 558 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47463 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151416 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14870 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14870 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14870 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18932 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27117 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68365 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13564 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84062 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90131 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43979 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37749 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73539 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58999 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10912434 - -
          Affymetrix Rat Gene 1.0 ST Array 10912438 - -
          Affymetrix Rat Gene 1.0 ST Array 10912436 - -
          Affymetrix Rat Gene 1.0 ST Array 10912437 - -
          Affymetrix Rat Gene 1.0 ST Array 10912435 - -
          Affymetrix Rat Gene 1.1 ST Array 10912433 - -
          Rat Genome 230 2.0 Array 1374025_at - -
          Rat Genome 230 2.0 Array 1374025_PM_at - -
          Affymetrix HT Rat Focus Array 1374025_PM_at - -
          Affymetrix HT Rat Focus Array 1374025_at - -
          Affymetrix Rat Clariom S Assay PSR0800009879.rn.1 TC0800001354.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800009881.rn.1 TC0800001354.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800009882.rn.1 TC0800001354.rn.1 RTA TC
          Rat Expression Set 230 1374025_PM_at - -
          Rat Expression Set 230 1374025_at - -
          Affymetrix Rat Gene 1.0 ST Array 10912433 - -
          Rat Genome U34 Set rc_AA963282_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20834297 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline