Search
Search
| Gene Symbol: Dgcr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107080.2 | NP_001100550.1 | 2 - 3 | 289 | 57 | |
| XM_006248609.2 | XP_006248671.1 | 2 - 3 | 318 | 57 | ||
| GenBank mRNA | BC158828.1 | - | 2 - 3 | 289 | 7378165 | 57 |
Set Membership: |
|