Search
Search

Ccdc116| Gene Symbol: Ccdc116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024740.1 | NP_001019911.1 | 2 - 3 | 251 | 62 | |
| XM_006248676.2 | XP_006248738.1 | 2 - 3 | 373 | 62 | ||
| GenBank mRNA | BC097459.1 | - | 2 - 3 | 251 | 7457223 | 62 |
Set Membership: |
|