Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008335.1 | NP_001008336.1 | 5 - 6 | 535 | 123 | |
XM_006248534.2 | XP_006248596.1 | 5 - 6 | 551 | 123 | ||
XM_006248535.2 | XP_006248597.1 | 5 - 6 | 551 | 123 | ||
GenBank mRNA | BC085859.1 | - | 5 - 6 | 535 | 7190333 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |