Search
Search
| Gene Symbol: Chrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057134.1 | NP_476475.1 | 14 - 15 | 1830 | 64 | |
| XM_006248556.2 | XP_006248618.1 | 14 - 15 | 2212 | 64 | ||
| XM_006248557.2 | XP_006248619.1 | 14 - 15 | 2236 | 64 | ||
| GenBank mRNA | FQ222446.1 | - | 14 - 15 | 1869 | 64 | |
| FQ223218.1 | - | 14 - 15 | 1869 | 64 | ||
Set Membership: |
|