Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01405094_g1
          See other TRMT2A GE Assays ›
          Gene Symbol:
          Trmt2a
          Gene Name:
          tRNA methyltransferase 2 homolog A (S. cerevisiae)
          Gene Aliases:
          Htf9c
          Chromosome Location:
          Chr.11: 86885671 - 86890489 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01405094_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trmt2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011895.1 NP_001011895.1 9 - 10 1648 63
          XM_006248604.1 XP_006248666.1 9 - 10 1860 63
          XM_006248605.1 XP_006248667.1 9 - 10 1619 63
          GenBank mRNA BC083619.1 - 9 - 10 1648 7133938 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001988.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trmt2a

          Panther Classification:

          Molecular Function -

          RNA methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA methylation
          RNA processing
          nucleotide binding
          RNA methyltransferase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P995959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641241 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P995959 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P995959 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P995959 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P995959 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755611 - -
          Affymetrix Rat Gene 1.0 ST Array 10755615 - -
          Affymetrix Rat Gene 1.0 ST Array 10755613 - -
          Affymetrix Rat Gene 1.0 ST Array 10755609 - -
          Affymetrix Rat Gene 1.0 ST Array 10755605 - -
          Affymetrix Rat Gene 1.0 ST Array 10755618 - -
          Affymetrix Rat Gene 1.0 ST Array 10755608 - -
          Affymetrix Rat Gene 1.0 ST Array 10755607 - -
          Affymetrix Rat Gene 1.0 ST Array 10755612 - -
          Affymetrix Rat Gene 1.0 ST Array 10755614 - -
          Affymetrix Rat Gene 1.0 ST Array 10755610 - -
          Affymetrix Rat Gene 1.0 ST Array 10755606 - -
          Affymetrix Rat Gene 1.0 ST Array 10755616 - -
          Affymetrix Rat Gene 1.1 ST Array 10755604 - -
          Rat Genome 230 2.0 Array 1395870_PM_at - -
          Rat Genome 230 2.0 Array 1395870_at - -
          Rat Genome 230 2.0 Array 1383125_at - -
          Rat Genome 230 2.0 Array 1383125_PM_at - -
          Affymetrix HT Rat Focus Array 1383125_PM_at - -
          Affymetrix HT Rat Focus Array 1383125_at - -
          Affymetrix HT Rat Focus Array 1395870_at - -
          Affymetrix HT Rat Focus Array 1395870_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100013075.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013063.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013068.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013064.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013081.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013079.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013066.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013069.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013073.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013076.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013077.rn.1 TC1100001988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013071.rn.1 TC1100001988.rn.1 RTA TC
          Rat Expression Set 230 1383125_at - -
          Rat Expression Set 230 1395870_PM_at - -
          Rat Expression Set 230 1395870_at - -
          Rat Expression Set 230 1383125_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755604 - -
          Rat Genome U34 Set rc_AI012598_at - -
          Rat Genome U34 Set rc_AI169719_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22304340 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376400 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline