Search
Search
| Gene Symbol: Cdc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105866.2 | NP_001099336.1 | 8 - 9 | 677 | 61 | |
| XM_008768866.1 | XP_008767088.1 | 9 - 10 | 999 | 61 | ||
| GenBank mRNA | BC161946.1 | - | 8 - 9 | 677 | 9025372 | 61 |
Set Membership: |
|