Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01405131_m1
          See other TBCCD1 GE Assays ›
          Gene Symbol:
          Tbccd1
          Gene Name:
          TBCC domain containing 1
          Gene Aliases:
          Crygs
          Chromosome Location:
          Chr.11: 81757966 - 81794367 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01405131_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbccd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012016.1 NP_001012016.1 7 - 8 1820 67
          XM_006248545.2 XP_006248607.1 6 - 7 1794 67
          XM_006248546.2 XP_006248608.1 6 - 7 1942 67
          GenBank mRNA BC089823.1 - 7 - 8 1820 7133137 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000887.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tbccd1

          Gene Ontology Categories:

          Function(s)

          cell morphogenesis
          regulation of cell shape
          regulation of cell migration
          maintenance of centrosome location
          maintenance of Golgi location

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052110 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P085467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500350 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149346 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085467 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052110 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085467 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052110 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052110 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P085467 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103541 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14733 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74749 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751998 - -
          Affymetrix Rat Gene 1.0 ST Array 10751997 - -
          Affymetrix Rat Gene 1.0 ST Array 10752003 - -
          Affymetrix Rat Gene 1.0 ST Array 10751995 - -
          Affymetrix Rat Gene 1.0 ST Array 10752002 - -
          Affymetrix Rat Gene 1.0 ST Array 10751999 - -
          Affymetrix Rat Gene 1.0 ST Array 10752001 - -
          Affymetrix Rat Gene 1.0 ST Array 10751996 - -
          Affymetrix Rat Gene 1.0 ST Array 10752000 - -
          Affymetrix Rat Gene 1.0 ST Array 10751994 - -
          Affymetrix Rat Gene 1.1 ST Array 10751993 - -
          Rat Genome 230 2.0 Array 1382349_PM_at - -
          Rat Genome 230 2.0 Array 1382349_at - -
          Rat Genome 230 2.0 Array 1388435_PM_at - -
          Rat Genome 230 2.0 Array 1388435_at - -
          Affymetrix HT Rat Focus Array 1382349_PM_at - -
          Affymetrix HT Rat Focus Array 1382349_at - -
          Affymetrix Rat Clariom S Assay PSR1100004924.rn.1 TC1100000887.rn.1 RTA TC
          Rat Expression Set 230 1382349_PM_at - -
          Rat Expression Set 230 1382349_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751993 - -
          Rat Genome U34 Set rc_AA957950_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21522083 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376644 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline