Search
Search
| Gene Symbol: Zdhhc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039021.1 | NP_001034110.1 | 9 - 10 | 1249 | 66 | |
| XM_006248610.2 | XP_006248672.1 | 9 - 10 | 1248 | 66 | ||
| GenBank mRNA | AY871204.1 | - | 9 - 10 | 1249 | 66 | |
Set Membership: |
|