Search
Search
| Gene Symbol: Dgcr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012472.1 | NP_001012490.1 | 1 - 2 | 153 | 72 | |
| XM_006248635.2 | XP_006248697.1 | 1 - 2 | 201 | 72 | ||
| XM_006248636.2 | XP_006248698.1 | 1 - 2 | 201 | 72 | ||
| GenBank mRNA | BC089898.1 | - | 1 - 2 | 153 | 7382217 | 72 |
Set Membership: |
|