Search
Search
| Gene Symbol: Liph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044279.1 | NP_001037744.1 | 1 - 2 | 131 | 67 | |
| GenBank mRNA | BC062045.1 | - | 137 | 67 | ||
| BC107932.1 | - | 1 - 2 | 131 | 7124552 | 67 | |
Set Membership: |
|