Search
Search
| Gene Symbol: Vps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248575.2 | XP_006248637.1 | 31 - 32 | 2881 | 54 | |
| XM_006248577.2 | XP_006248639.1 | 31 - 32 | 2873 | 54 | ||
| XM_006248578.2 | XP_006248640.1 | 31 - 32 | 2905 | 54 | ||
| XM_006248579.2 | XP_006248641.1 | 30 - 31 | 2875 | 54 | ||
| XM_008768830.1 | XP_008767052.1 | 31 - 32 | 3080 | 54 | ||
| XM_008768831.1 | XP_008767053.1 | 31 - 32 | 2881 | 54 | ||
Set Membership: |
|