Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01405759_m1
          See other NDFIP1 GE Assays ›
          Gene Symbol:
          Ndfip1
          Gene Name:
          Nedd4 family interacting protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 31574753 - 31625286 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01405759_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndfip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013059.1 NP_001013077.1 6 - 7 699 82
          XM_006254633.2 XP_006254695.1 6 - 7 767 82
          GenBank mRNA BC085887.1 - 6 - 7 699 7192378 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000374.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndfip1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of myeloid leukocyte differentiation
          negative regulation of type 2 immune response
          cellular iron ion homeostasis
          signal transduction
          negative regulation of gene expression
          positive regulation of protein ubiquitination
          negative regulation of transporter activity
          negative regulation of interleukin-4 production
          negative regulation of T cell proliferation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          regulation of lymphocyte differentiation
          positive regulation of protein catabolic process
          negative regulation of isotype switching to IgE isotypes
          regulation of isotype switching to IgG isotypes
          negative regulation of inflammatory response
          negative regulation of protein transport
          signal transducer activity
          protein binding
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P166216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68804 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P166216 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P166216 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P166216 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P166216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88128 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37289 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12237 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10801294 - -
          Affymetrix Rat Gene 1.0 ST Array 10801291 - -
          Affymetrix Rat Gene 1.0 ST Array 10801296 - -
          Affymetrix Rat Gene 1.0 ST Array 10801295 - -
          Affymetrix Rat Gene 1.0 ST Array 10801297 - -
          Affymetrix Rat Gene 1.0 ST Array 10801292 - -
          Affymetrix Rat Gene 1.0 ST Array 10801293 - -
          Affymetrix Rat Gene 1.1 ST Array 10801290 - -
          Rat Genome 230 2.0 Array 1398893_at - -
          Rat Genome 230 2.0 Array 1398893_PM_at - -
          Affymetrix HT Rat Focus Array 1398893_PM_at - -
          Affymetrix HT Rat Focus Array 1398893_at - -
          Affymetrix Rat Clariom S Assay PSR1800002877.rn.1 TC1800000374.rn.1 RTA TC
          Rat Expression Set 230 1398893_at - -
          Rat Expression Set 230 1398893_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10801290 - -
          Rat Genome U34 Set rc_AI008958_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20802286 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650482 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline