Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01405921_m1
          See other SPATA24 GE Assays ›
          Gene Symbol:
          Spata24
          Gene Name:
          spermatogenesis associated 24
          Gene Aliases:
          RGD1311742
          Chromosome Location:
          Chr.18: 28446592 - 28454725 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01405921_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spata24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025636.2 NP_001020807.2 1 - 2 127 109
          XM_006254545.2 XP_006254607.1 1 - 2 246 109
          GenBank mRNA DQ080016.1 - 1 - 2 127 109

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the DQ080016.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spata24

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          multicellular organism development
          spermatogenesis
          cell differentiation
          DNA binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P513490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734962 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P513490 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P513490 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P513490 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P513490 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41805 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803908 - -
          Affymetrix Rat Gene 1.0 ST Array 10803906 - -
          Affymetrix Rat Gene 1.0 ST Array 10803907 - -
          Affymetrix Rat Gene 1.0 ST Array 10803909 - -
          Affymetrix Rat Gene 1.0 ST Array 10803910 - -
          Affymetrix Rat Gene 1.1 ST Array 10803903 - -
          Rat Genome 230 2.0 Array 1375553_PM_at - -
          Rat Genome 230 2.0 Array 1398444_PM_at - -
          Rat Genome 230 2.0 Array 1375553_at - -
          Rat Genome 230 2.0 Array 1398444_at - -
          Affymetrix HT Rat Focus Array 1375553_at - -
          Affymetrix HT Rat Focus Array 1398444_PM_at - -
          Affymetrix HT Rat Focus Array 1375553_PM_at - -
          Affymetrix HT Rat Focus Array 1398444_at - -
          Affymetrix Rat Clariom S Assay PSR1800008900.rn.1 TC1800001249.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800008901.rn.1 TC1800001249.rn.1 RTA TC
          Rat Expression Set 230 1375553_at - -
          Rat Expression Set 230 1398444_PM_at - -
          Rat Expression Set 230 1375553_PM_at - -
          Rat Expression Set 230 1398444_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803903 - -
          Rat Genome U34 Set rc_AI111844_at - -
          Rat Genome U34 Set rc_AI112160_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22370228 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357778 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline