Search
Search
| Gene Symbol: Wdr33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107398.2 | NP_001100868.2 | 2 - 3 | 368 | 77 | |
| XM_006254589.2 | XP_006254651.1 | 2 - 3 | 435 | 77 | ||
| XM_008772041.1 | XP_008770263.1 | 2 - 3 | 417 | 77 | ||
Set Membership: |
|