Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01407113_m1
          See other ETS2 GE Assays ›
          Gene Symbol:
          Ets2
          Gene Name:
          v-ets avian erythroblastosis virus E26 oncogene homolog 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 36075709 - 36092495 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01407113_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ets2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107107.1 NP_001100577.2 8 - 9 1298 62
          XM_006248177.2 XP_006248239.1 8 - 9 1516 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000340.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          mesoderm development
          cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          primitive streak formation
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          glucocorticoid receptor binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P761087 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P761087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078108 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P761087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078108 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078108 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P761087 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P761087 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P267799 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750466 - -
          Affymetrix Rat Gene 1.0 ST Array 10750464 - -
          Affymetrix Rat Gene 1.0 ST Array 10750469 - -
          Affymetrix Rat Gene 1.0 ST Array 10750463 - -
          Affymetrix Rat Gene 1.0 ST Array 10750462 - -
          Affymetrix Rat Gene 1.0 ST Array 10750465 - -
          Affymetrix Rat Gene 1.0 ST Array 10750467 - -
          Affymetrix Rat Gene 1.0 ST Array 10750461 - -
          Affymetrix Rat Gene 1.0 ST Array 10750468 - -
          Affymetrix Rat Gene 1.1 ST Array 10750460 - -
          Rat Genome 230 2.0 Array 1372564_at - -
          Rat Genome 230 2.0 Array 1372564_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100001661.rn.1 TC1100000340.rn.1 RTA TC
          Rat Expression Set 230 1372564_PM_at - -
          Rat Expression Set 230 1372564_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750460 - -
          Rat Genome U34 Set rc_AA851620_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21725547 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363372 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline