Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01407364_g1
          See other HDAC10 GE Assays ›
          Gene Symbol:
          Hdac10
          Gene Name:
          histone deacetylase 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 130102335 - 130108346 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01407364_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hdac10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001035000.1 NP_001030172.1 17 - 18 1978 56
          XM_006242218.1 XP_006242280.1 19 - 20 3121 56
          XM_006242219.1 XP_006242281.1 18 - 19 3064 56
          XM_006242220.1 XP_006242282.1 18 - 19 2944 56
          GenBank mRNA BC092573.1 - 17 - 18 1978 7323327 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003686.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein deacetylation
          oligodendrocyte development
          histone deacetylation
          negative regulation of transcription, DNA-templated
          histone H3-K9 modification
          histone H3 deacetylation
          histone H4 deacetylation
          molecular_function
          histone deacetylase activity
          enzyme binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          protein deacetylase activity
          histone deacetylase binding
          NAD-dependent histone deacetylase activity (H3-K9 specific)
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          NAD-dependent histone deacetylase activity (H3-K18 specific)

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P121935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100142 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121935 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121935 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P121935 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121935 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10906138 - -
          Affymetrix Rat Gene 1.0 ST Array 10906123 - -
          Affymetrix Rat Gene 1.0 ST Array 10906131 - -
          Affymetrix Rat Gene 1.0 ST Array 10906127 - -
          Affymetrix Rat Gene 1.0 ST Array 10906132 - -
          Affymetrix Rat Gene 1.0 ST Array 10906135 - -
          Affymetrix Rat Gene 1.0 ST Array 10906136 - -
          Affymetrix Rat Gene 1.0 ST Array 10906122 - -
          Affymetrix Rat Gene 1.0 ST Array 10906133 - -
          Affymetrix Rat Gene 1.0 ST Array 10906134 - -
          Affymetrix Rat Gene 1.0 ST Array 10906129 - -
          Affymetrix Rat Gene 1.0 ST Array 10906125 - -
          Affymetrix Rat Gene 1.0 ST Array 10906121 - -
          Affymetrix Rat Gene 1.0 ST Array 10906141 - -
          Affymetrix Rat Gene 1.0 ST Array 10906139 - -
          Affymetrix Rat Gene 1.0 ST Array 10906140 - -
          Affymetrix Rat Gene 1.0 ST Array 10906137 - -
          Affymetrix Rat Gene 1.0 ST Array 10906126 - -
          Affymetrix Rat Gene 1.0 ST Array 10906120 - -
          Affymetrix Rat Gene 1.1 ST Array 10906119 - -
          Rat Genome 230 2.0 Array 1384991_PM_a_at - -
          Rat Genome 230 2.0 Array 1390700_PM_s_at - -
          Rat Genome 230 2.0 Array 1384992_PM_at - -
          Rat Genome 230 2.0 Array 1380608_at - -
          Rat Genome 230 2.0 Array 1384991_a_at - -
          Rat Genome 230 2.0 Array 1384992_at - -
          Rat Genome 230 2.0 Array 1380608_PM_at - -
          Rat Genome 230 2.0 Array 1390700_s_at - -
          Affymetrix HT Rat Focus Array 1390700_s_at - -
          Affymetrix HT Rat Focus Array 1384991_PM_a_at - -
          Affymetrix HT Rat Focus Array 1384992_at - -
          Affymetrix HT Rat Focus Array 1384991_a_at - -
          Affymetrix HT Rat Focus Array 1380608_PM_at - -
          Affymetrix HT Rat Focus Array 1384992_PM_at - -
          Affymetrix HT Rat Focus Array 1390700_PM_s_at - -
          Affymetrix HT Rat Focus Array 1380608_at - -
          Affymetrix Rat Clariom S Assay PSR0700029147.rn.1 TC0700003686.rn.1 RTA TC
          Rat Expression Set 230 1390700_s_at - -
          Rat Expression Set 230 1390700_PM_s_at - -
          Rat Expression Set 230 1384992_at - -
          Rat Expression Set 230 1384991_a_at - -
          Rat Expression Set 230 1384991_PM_a_at - -
          Rat Expression Set 230 1384992_PM_at - -
          Rat Expression Set 230 1380608_PM_at - -
          Rat Expression Set 230 1380608_at - -
          Affymetrix Rat Gene 1.0 ST Array 10906119 - -
          Rat Genome U34 Set rc_AI236142_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22131183 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline