Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01407714_m1
          See other LRRK2 GE Assays ›
          Gene Symbol:
          Lrrk2
          Gene Name:
          leucine-rich repeat kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 132857313 - 133018549 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01407714_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191789.1 NP_001178718.1 47 - 48 7026 68
          XM_006242190.2 XP_006242252.1 47 - 48 7421 68
          XM_008765689.1 XP_008763911.1 46 - 47 7274 68
          GenBank mRNA BC091422.1 - 3 - 4 409 68
          BC168217.1 - 4 - 5 586 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001669.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          activation of MAPK activity
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          protein phosphorylation
          endocytosis
          response to oxidative stress
          small GTPase mediated signal transduction
          neuromuscular junction development
          determination of adult lifespan
          positive regulation of autophagy
          regulation of protein kinase A signaling
          negative regulation of protein processing
          regulation of neuron maturation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          olfactory bulb development
          tangential migration from the subventricular zone to the olfactory bulb
          positive regulation of protein ubiquitination
          negative regulation of protein binding
          positive regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of GTPase activity
          cellular response to oxidative stress
          cellular protein localization
          intracellular signal transduction
          regulation of kidney size
          exploration behavior
          locomotory exploration behavior
          regulation of locomotion
          regulation of membrane potential
          positive regulation of programmed cell death
          positive regulation of GTPase activity
          protein autophosphorylation
          intracellular distribution of mitochondria
          neuron projection morphogenesis
          regulation of mitochondrial depolarization
          regulation of synaptic transmission, glutamatergic
          excitatory postsynaptic potential
          regulation of dopamine receptor signaling pathway
          positive regulation of dopamine receptor signaling pathway
          regulation of dendritic spine morphogenesis
          neuron death
          cellular response to manganese ion
          cellular response to organic cyclic compound
          reactive oxygen species metabolic process
          regulation of neuron death
          negative regulation of neuron death
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          positive regulation of protein autoubiquitination
          regulation of neuroblast proliferation
          negative regulation of thioredoxin peroxidase activity by peptidyl-threonine phosphorylation
          negative regulation of hydrogen peroxide-induced cell death
          negative regulation of protein targeting to mitochondrion
          cellular response to dopamine
          regulation of branching morphogenesis of a nerve
          regulation of synaptic vesicle exocytosis
          negative regulation of peroxidase activity
          SNARE binding
          glycoprotein binding
          molecular_function
          actin binding
          GTPase activity
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase activity
          GTPase activator activity
          ATP binding
          GTP binding
          tubulin binding
          kinase activity
          Rho GTPase binding
          syntaxin-1 binding
          clathrin binding
          GTP-dependent protein kinase activity
          peroxidase inhibitor activity
          identical protein binding
          protein homodimerization activity
          ion channel binding
          protein kinase A binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P520848 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P520848 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P520848 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P520848 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P520848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898721 - -
          Affymetrix Rat Gene 1.0 ST Array 10898725 - -
          Affymetrix Rat Gene 1.0 ST Array 10898720 - -
          Affymetrix Rat Gene 1.0 ST Array 10898727 - -
          Affymetrix Rat Gene 1.0 ST Array 10898723 - -
          Affymetrix Rat Gene 1.0 ST Array 10898724 - -
          Affymetrix Rat Gene 1.0 ST Array 10898722 - -
          Affymetrix Rat Gene 1.0 ST Array 10898726 - -
          Affymetrix Rat Gene 1.1 ST Array 10898676 - -
          Affymetrix Rat Clariom S Assay PSR0700012867.rn.1 TC0700001669.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10898676 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374948 - -
          RatRef-12 Expression BeadChip ILMN_50629 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline