Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lama4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751975.3 | XP_003752023.1 | 33 - 34 | 5423 | 59 | |
GenBank mRNA | FQ221747.1 | - | 33 - 34 | 5105 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |