Search
Search
| Gene Symbol: Nt5dc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106393.1 | NP_001099863.1 | 1 - 2 | 199 | 68 | |
| XM_008772980.1 | XP_008771202.1 | 1 - 2 | 215 | 68 | ||
| XM_008772981.1 | XP_008771203.1 | 1 - 2 | 215 | 68 | ||
| GenBank mRNA | BC162021.1 | - | 1 - 2 | 163 | 9098589 | 68 |
Set Membership: |
|