Search
Search
| Gene Symbol: Wdr37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107362.1 | NP_001100832.1 | 3 - 4 | 494 | 94 | |
| XM_008771733.1 | XP_008769955.1 | 2 - 3 | 843 | 94 | ||
| XM_008771734.1 | XP_008769956.1 | 2 - 3 | 363 | 94 | ||
Set Membership: |
|