Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01408132_m1
          See other CYP4F40 GE Assays ›
          Gene Symbol:
          Cyp4f40
          Gene Name:
          cytochrome P450, family 4, subfamily f, polypeptide 40
          Gene Aliases:
          RGD1559596
          Chromosome Location:
          Chr.7: 14865941 - 14880084 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01408132_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp4f40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109360.1 NP_001102830.1 1 - 2 200 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000276.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          menaquinone catabolic process
          phylloquinone catabolic process
          vitamin K catabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
          heme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133600 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078255 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078255 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078255 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17754 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894196 - -
          Affymetrix Rat Gene 1.0 ST Array 10894199 - -
          Affymetrix Rat Gene 1.0 ST Array 10894197 - -
          Affymetrix Rat Gene 1.0 ST Array 10894195 - -
          Affymetrix Rat Gene 1.0 ST Array 10894192 - -
          Affymetrix Rat Gene 1.0 ST Array 10894189 - -
          Affymetrix Rat Gene 1.0 ST Array 10894194 - -
          Affymetrix Rat Gene 1.0 ST Array 10894198 - -
          Affymetrix Rat Gene 1.0 ST Array 10894191 - -
          Affymetrix Rat Gene 1.0 ST Array 10894193 - -
          Affymetrix Rat Gene 1.0 ST Array 10894190 - -
          Affymetrix Rat Gene 1.1 ST Array 10894188 - -
          Affymetrix Rat Clariom S Assay PSR0700002611.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002615.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002616.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002617.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002610.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002618.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002620.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002612.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002621.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002613.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002614.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002619.rn.1 TC0700000276.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10894188 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22341430 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650601 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline