Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01408683_m1
          See other EPB4.1L5 GE Assays ›
          Gene Symbol:
          Epb4.1l5
          Gene Name:
          erythrocyte membrane protein band 4.1-like 5
          Gene Aliases:
          Epb41l5
          Chromosome Location:
          Chr.13: 35622472 - 35673449 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01408683_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Epb4.1l5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012023.1 NP_001012023.1 10 - 11 942 140
          GenBank mRNA BC090012.1 - 10 - 11 942 7324591 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001260.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Epb4.1l5

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis involved in differentiation
          in utero embryonic development
          somitogenesis
          epithelial to mesenchymal transition
          neural plate morphogenesis
          positive regulation of cell-matrix adhesion
          epithelial cell morphogenesis
          apical constriction
          substrate-dependent cell migration, cell attachment to substrate
          ectoderm development
          endoderm development
          mesoderm development
          mesoderm migration involved in gastrulation
          unidimensional cell growth
          posttranscriptional regulation of gene expression
          positive regulation of epithelial cell migration
          positive regulation of epithelial to mesenchymal transition
          negative regulation of cell-cell adhesion
          actin cytoskeleton organization
          actomyosin structure organization
          negative regulation of protein binding
          positive regulation of protein binding
          somite rostral/caudal axis specification
          axial mesoderm development
          axial mesoderm morphogenesis
          paraxial mesoderm development
          embryonic foregut morphogenesis
          positive regulation of focal adhesion assembly
          regulation of establishment of protein localization
          left/right axis specification
          cellular response to transforming growth factor beta stimulus
          cytoskeletal protein binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109230 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49673 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159101 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80298 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014371 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014371 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014371 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P715347 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103801 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80341 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767127 - -
          Affymetrix Rat Gene 1.0 ST Array 10767114 - -
          Affymetrix Rat Gene 1.0 ST Array 10767124 - -
          Affymetrix Rat Gene 1.0 ST Array 10767117 - -
          Affymetrix Rat Gene 1.0 ST Array 10767118 - -
          Affymetrix Rat Gene 1.0 ST Array 10767125 - -
          Affymetrix Rat Gene 1.0 ST Array 10767116 - -
          Affymetrix Rat Gene 1.0 ST Array 10767126 - -
          Affymetrix Rat Gene 1.0 ST Array 10767128 - -
          Affymetrix Rat Gene 1.0 ST Array 10767119 - -
          Affymetrix Rat Gene 1.0 ST Array 10767123 - -
          Affymetrix Rat Gene 1.0 ST Array 10767112 - -
          Affymetrix Rat Gene 1.0 ST Array 10767129 - -
          Affymetrix Rat Gene 1.0 ST Array 10767113 - -
          Affymetrix Rat Gene 1.0 ST Array 10767120 - -
          Affymetrix Rat Gene 1.0 ST Array 10767121 - -
          Affymetrix Rat Gene 1.0 ST Array 10767115 - -
          Affymetrix Rat Gene 1.0 ST Array 10767122 - -
          Affymetrix Rat Gene 1.1 ST Array 10767102 - -
          Rat Genome 230 2.0 Array 1374984_at - -
          Rat Genome 230 2.0 Array 1374984_PM_at - -
          Affymetrix HT Rat Focus Array 1374984_at - -
          Affymetrix HT Rat Focus Array 1374984_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300008338.rn.1 TC1300001260.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008337.rn.1 TC1300001260.rn.1 RTA TC
          Rat Expression Set 230 1374984_PM_at - -
          Rat Expression Set 230 1374984_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767102 - -
          Rat Genome U34 Set rc_AA858925_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline