Search
Search
| Gene Symbol: Pdcd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107452.1 | NP_001100922.1 | 2 - 3 | 291 | 113 | |
| XM_006227784.2 | XP_006227846.1 | 2 - 3 | 280 | 113 | ||
| GenBank mRNA | BC168744.1 | - | 2 - 3 | 248 | 6920123 | 113 |
| FQ213638.1 | - | 2 - 3 | 212 | 113 | ||
| FQ219953.1 | - | 2 - 3 | 212 | 113 | ||
| FQ219997.1 | - | 2 - 3 | 212 | 113 | ||
| FQ231349.1 | - | 2 - 3 | 212 | 113 | ||
| FQ231760.1 | - | 2 - 3 | 212 | 113 | ||
| FQ234686.1 | - | 2 - 3 | 212 | 113 | ||
Set Membership: |
|