Search
Search
| Gene Symbol: Herc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107520.1 | NP_001100990.1 | 87 - 88 | 13614 | 56 | |
| XM_006229300.2 | XP_006229362.1 | 88 - 89 | 13802 | 56 | ||
| XM_006229301.2 | XP_006229363.1 | 88 - 89 | 13830 | 56 | ||
| XM_006229302.2 | XP_006229364.1 | 88 - 89 | 13787 | 56 | ||
| XM_006229304.2 | XP_006229366.1 | 87 - 88 | 13694 | 56 | ||
Set Membership: |
|