Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01409992_m1
          See other FKBP14 GE Assays ›
          Gene Symbol:
          Fkbp14
          Gene Name:
          FK506 binding protein 14
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 84753628 - 84768249 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01409992_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fkbp14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013210.1 NP_001013228.1 2 - 3 460 83
          GenBank mRNA BC083746.1 - 2 - 3 460 7192246 83
          FQ216652.1 - 2 - 3 507 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fkbp14

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          calcium ion binding
          FK506 binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P326233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69126 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158856 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P326233 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P326233 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P326233 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P326233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12724 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93884 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862647 - -
          Affymetrix Rat Gene 1.0 ST Array 10862645 - -
          Affymetrix Rat Gene 1.0 ST Array 10862644 - -
          Affymetrix Rat Gene 1.0 ST Array 10862648 - -
          Affymetrix Rat Gene 1.1 ST Array 10862643 - -
          Rat Genome 230 2.0 Array 1383976_at - -
          Rat Genome 230 2.0 Array 1383976_PM_at - -
          Rat Genome 230 2.0 Array 1372525_PM_at - -
          Rat Genome 230 2.0 Array 1372525_at - -
          Affymetrix HT Rat Focus Array 1372525_PM_at - -
          Affymetrix HT Rat Focus Array 1372525_at - -
          Affymetrix Rat Clariom S Assay PSR0400020957.rn.1 TC0400002940.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020954.rn.1 TC0400002940.rn.1 RTA TC
          Rat Expression Set 230 1372525_PM_at - -
          Rat Expression Set 230 1372525_at - -
          Affymetrix Rat Gene 1.0 ST Array 10862643 - -
          Rat Genome U34 Set rc_AA848418_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline